Learn More
Trajan™ CTC PAL Removable Needle Syringe
Click to view available options
Volume (Metric):
250 μL
Unit Size:
Each
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for High performance liquid chromatology (HPLC)
Specifications
Specifications
| Material | PTFE (Plunger) |
| Needle Gauge | 22 G |
| Diameter (Metric) Inner Needle | 0.37 mm |
| Diameter (Metric) Outer Needle | 0.72 mm |
| Product Type | HPLC Autosampler Syringe |
| Volume (Metric) | 250 μL |
| Needle Point Style | LC |