missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SF 11

Product Code. 18005922
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18005922 10 mg 10mg
18064522 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18005922 Supplier Tocris Bioscience™ Supplier No. 3898/10
  • ONLINE EXCLUSIVE PRICE
    306.00€
    288.75€ / 10mg
    Save 17.25€
    5% Off
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

NPY Y2 receptor antagonist

SF 11 is an NPY Y2 antagonist (IC50 = 199 nM). Displays no affinity for Y1 receptor at concentrations up to 35 μM. Causes significant lethality in mice. Displays antidepressant effects in rats. Brain penetrant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 443292-81-7
Molecular Formula C27H30N2O2S

Specifications

Quantity 10 mg
Formula Weight 446.6
Purity >99%
Molecular Weight (g/mol) 446.609
Chemical Name or Material SF 11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.