Learn More
VACUUBRAND™ Service Kits for Oil-Free Diaphragm Vacuum Pumps
Click to view available options
For Use With:
MD1C, PC3001 Diaphragm Pump
ME 8 NT
ME1, ME1C Diaphragm Pump
ME2C NT Diaphragm Pump
MZ 2 NT, ME 4 NT + VARIO
MZ 4C-MD 4C
MZ2D NT VARIO
Unit Size:
1 set
Each
Description
- Includes appropriate diaphragms, valves, and sealing rings, if appropriate
- Does not include spanner wrench
Specifications
Specifications
| For Use With | MD1C, PC3001 Diaphragm Pump |
| Type | Service Kit |