Learn More
Abeba™ Service 3000 Shoes
Click to view available options
Size:
34
35
36
37
38
39
40
41
42
Unit Size:
1 pair
Description
- CE
- Smooth leather
- Black
- Slip resistance outsole sole
- Sanitized treated
- Cushioned ankle supports
- Instep area with lacing
- Heel 45 mm
- Comfort foodbed
Specifications
Specifications
| Certifications/Compliance | CE |
| Color | Black |
| Material | Leather |
| Size | 42 |
| Sole Features | Slip-resistant |
| Style | 3000 |