missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ SERPING1 Recombinant Protein

Brand:  Abnova™ H00000710-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16102411

  • 508.00€ / 25µg

Estimated Shipment: 02-05-2023
to see stock.
Add to basket

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

Antibody Production, ELISA, Protein Array, Western Blotting (Recombinant Protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Human SERPING1 Full-length ORF Recombinant Protein with GST-tag at N-terminal
In vitro wheat germ expression system
12.5% SDS-PAGE stained with Coomassie Blue
Wheat Germ (in vitro)
Human
Solution
80.52
8
Glutathione Sepharose 4 Fast Flow
25μg
-80°C
Yes
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers