Learn More
Cytiva Sera-Mag SpeedBeads™ Streptavidin magnetic beads
Product Code. 11869912 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
100 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11869912 1 mL Each
11849912 5 mL Each
11859912 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11869912 Supplier Cytiva Supplier No. 66152104011150
SpeedBeads Streptavidin exhibit a high affinity when binding to biotinylated ligands such as proteins, nucleic acids and peptides. They respond much faster to a magnetic field to shorten assay times and improve precision and also move faster through viscous solutions.
  • Available with a 3500-4500 pmol/mg biotin-binding range
  • Their cauliflower-like structure gives Sera-Mag beads high binding capacity
  • Combine fast reaction kinetics and low, non-specific binding for increased throughput and precision in such immunoassay and molecular biology applications as sample preparation and assay development for genomics and proteomics
TRUSTED_SUSTAINABILITY
Specifications
Concentration 1% solids (10mg/mL)
Content And Storage 2°C to 8°C
For Use With (Application) Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets
Quantity 1 mL
Diameter (Metric) 1 μm