Description
- Available with a 3500-4500 pmol/mg biotin-binding range
- Their cauliflower-like structure gives Sera-Mag beads high binding capacity
- Combine fast reaction kinetics and low, non-specific binding for increased throughput and precision in such immunoassay and molecular biology applications as sample preparation and assay development for genomics and proteomics
Specifications
Specifications
| Concentration | 1% solids (10mg/mL) |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets |
| Quantity | 100 mL |
| Diameter (Metric) | 1 μm |