Learn More
Applied Biosystems™ SeqScape™ Software v4.0, Initial License
Product Code. 17404162 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 license
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17404162 1 license Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17404162 Supplier Applied Biosystems™ Supplier No. A38880
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
SeqScape Software is a resequencing package designed for mutation detection and analysis, SNP discovery and validation, pathogen sub-typing, allele identification, and sequence confirmation
  • This is an Initial License for SeqScape Software
  • SeqScape Software is a resequencing package designed for mutation detection and analysis, SNP discovery and validation, pathogen sub-typing, allele identification, and sequence confirmation
  • It provides library functions for comparison to a known group of sequences, as well as 21 CFR Part 11 functionalities
TRUSTED_SUSTAINABILITY
Specifications
Product Line SeqScape
Software Type Sequencing Data Analysis
Content And Storage Software/CD
Data Entry CE Sequencing (.ab1 files)
License Initial
Operating System Windows 10 Pro/IoT, Windows 7 Pro
Product Type SeqScape Software V4.0 Initial License
Quantity 1 license
Software Category Supports files from all AB Genetic Analyzers