Learn More
Hamilton™ Luer Lock Septum Adapter
Product Code. 10003354
Change view
Click to view available options
Connection Type:
Luer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connection Type Unit Size
10003354 Luer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10003354 Supplier Hamilton™ Supplier No. 31335
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
Turns any luer syringe into a sealed reaction container.
  • Samples can be added or removed from the sealed container using another syringe to pierce the replaceable septum
  • 1 adapter and 12 septa included
  • Sampling and storage applications
Specifications
For Use With (Application) Sampling and storage
Includes 12 replacement septa, 1 adapter
Product Type Septum Adapter
Connection Type Luer