Learn More
Macherey-Nagel™ Septa N 19 Polyethylene Caps
Click to view available options
Color:
Beige, Gray
Red-Orange
Material:
Polyethylene, Butyl, PTFE
Polyethylene, Natural Rubber, TEF
Unit Size:
Pack of 100
Description
- Septa N 19 for PE caps N 20
- Used as washer bottle closures or interim closures
- Packs of 100 closures each
Specifications
Specifications
| Color | Beige, Gray |
| Material | Polyethylene, Butyl, PTFE |
| Septum | Septa N 19 |
| Quantity | 100 Pk. |
| Thickness (Metric) | 1.3 mm |