Learn More
Cytiva Sephacryl™ High Resolution Media
Product Code. 11510794 Shop All Cytiva Products
Click to view available options
Quantity:
150 mL
750 mL
Product Type:
Sephacryl S-100 HR media
Sephacryl S-200 HR media
Sephacryl S-300 HR media
Sephacryl S-400 HR media
Flow Rate:
>125 cm/hr.
>150 cm/hr.
Particle Size:
25 to 75μm
50μm
Unit Size:
150mL
750mL
Product Code. 11510794
Brand: Cytiva 17060901
Purify proteins, polysaccharides, and other macromolecules by gel filtration at laboratory and industrial scale. Cytiva Life Sciences™Sephacryl™ High Resolution Media enables fast and reproducible purifications.
  • Fast and reproducible purification of proteins, polysaccharides, and other macromolecules
  • Gel filtration media providing high resolution with fast and easy operation
  • Available in five different fractionation ranges, allows for reproducible separations over a wide molecular weight range
  • BioProcess medium supported for industrial applications and well-established in approved processes
  • Available in different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
  • Long-term physical, chemical stability and easy handling
  • Hydrophilic nature enables maximum recovery and minimum     nonspecific adsorption
  • Average particle Size: 50μm
  • Matrix: Allyl dextran and N, N'-methylenebisacrylamide
  • pH stability working range: 3-11
  • Shelf Life: 5years
TRUSTED_SUSTAINABILITY
Specifications
Buffer 0.2M NaOH, 0.1M HCl, 1M Acetic Acid, 8M Urea, 6M Guanidine HCl, 1% SDS, 20% Ethanol, 30% Propanol, 30% Aceto nitrile (tested at 40°C for 1 week), 0.5M NaOH (only for cleaning-in-place)
Product Type Sephacryl S-400 HR media
Quantity 750 mL
Height (English) 5.9 in.
Height (Metric) 15cm
Flow Rate >150 cm/hr.
Max. Pressure 1 bar
pH Range 3 to 11
Particle Size 50μm
Support Type Allyl Dextran and N, N'-methylenebisacrylamide
Content And Storage 4° to 30°C
For Use With (Application) Purification of Proteins, Polysaccharides and other macromolecules
Matrix Gel Filtration Media
Show More Show Less
compliance-icons
Product Identifier
  • Sephacryl S-400 High Resolution, 750 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol