Learn More
Lenz Laborglasinstrumente™ Separating Funnels with Glass Plug
Product Code. 10316221
Change view
Click to view available options
Capacity (Metric):
2000 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10316221 2000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10316221 Supplier Lenz Laborglasinstrumente™ Supplier No. 04030073
  • ONLINE EXCLUSIVE PRICE
    281.00€
    183.21€ / Pack of 1
    Save 97.79€
    34% Off
Estimated Shipment: 14-10-2026
Made from Borosilicate glass. Lenz Laborglasinstrumente™ Dropping Funnels with Glass Plug are fitted with a stopcock and are excellent for transferring fluids.
  • NS Socket and PE stopper
  • Without scale, with glass plug
  • Inscription label
  • Borosilicate glass
Specifications
Capacity (Metric) 2000 mL
Material Borosilicate Glass
Pore Size 6.0 mm bore
Quantity 1
Stopper Material Polyethylene