Learn More
Lenz Laborglasinstrumente™ Separating Funnel
Product Code. 13459966
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
Unit Size:
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
13459966 250 mL Pack of 2
15380885 500 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13459966 Supplier Lenz Laborglasinstrumente™ Supplier No. 04004149
  • ONLINE EXCLUSIVE PRICE
    159.00€
    90.28€ / Pack of 2
    Save 68.72€
    43% Off
Estimated Shipment: 09-12-2026
Constructed of borosilicate glass. Lenz Laborglasinstrumente™ Separating Funnel includes screwthread retaining device, NS socket and PE stopper with PTFE plug. Without scale.
Separating funnel constructed of borosilicate glass.
  • Includes screwthread retaining device
  • NS socket and PE stopper with PTFE plug.
  • Ungraduated
Specifications
Capacity (Metric) 250 mL
Material Borosilicate Glass
Quantity 1Each
Stopcock 4mm; Polytetrafluoroethylene
Stopper Material Polyethylene
Stopper Number 29/32
Product Type Separatory Funnel