Learn More
WTW™ SenTix™ Epoxy Combination pH Electrode
GREENER_CHOICE
Product Code. 10557901
Click to view available options
Material:
Epoxy
Unit Size:
Each
Product Code. 10557901
Brand: WTW™ 103636
Only null left
Add to basket
Add to basket
For use with WTW ProfiLine and inoLab™ pH meter series, this SenTix™ Epoxy Combination pH Electrode is Ideal for portable measurements as well as for routine measurements in-the-laboratory.
  • Combination pH electrode, gel filled with temperature compensation and 10-ft cable
  • For use with WTW ProfiLine and inoLab™ pH meter series
  • Ideal for portable measurements as well as for routine measurements in-the-laboratory
  • Includes gel electrolyte, DIN plug, banana plug, integrated temperature sensor SenTix™ 41-3, gel electrolyte, Built-in NTC (30kilohms) temperature probe, Fiber diaphragm, 3m fixed cable
  • Basic pH combination electrode type
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) WTW ProfiLine and inoLab™ pH Meter Series
Type Basic pH Combination Electrode
For Use With (Application) Ideal for portable measurements as well as for routine measurements in-the-laboratory
Includes Gel electrolyte, DIN plug, banana plug, integrated temperature sensor SenTix™ 41-3, gel electrolyte, Built-in NTC (30kilohms) temperature probe, Fiber diaphragm, 3m fixed cable
Internal Reference Gel
Material Epoxy
pH Range 0 to 14
Length (Metric) 3 m
Length (Metric) Cable 3 m