Learn More
WTW™ SenTix™ 980-P pH Electrode
Click to view available options
Length (Metric):
120 mm
Unit Size:
Each
Description
- Digital wireless IDS pH combination electrode for precise, general lab measurements
- Liquid electrolyte, glass shaft, platinum wire junction and integrated temperature sensor
- Reliably measured values due to signal processing in the sensor
- Easy electrode replacement thanks to calibration data stored inside sensor
- Sensor quality monitoring via integrated QSC function
Specifications
Specifications
| Junction | Platinum Diaphragm |
| Material | Glass |
| pH Range | 0 to 14 |
| For Use With (Application) | Precise IDS pH Measurement in The Laboratory |
| Includes | Plug head for AS/IDS-x cables or wireless modules IDS WLM-S |
| Internal Reference | KCl 3 mol/L Ag+ free |
| Membrane | Cone |
| Temperature (Metric) Range | 0°C to 100°C |
| Length (Metric) | 120 mm |
| Diameter (Metric) | 12 mm |