Learn More
WTW™ SenTix™ 940-P pH Electrode
Click to view available options
pH Range:
0 to 14
Unit Size:
Each
Description
- Low maintenance IDS pH combination electrode with gel electrolyte, plastic shaft and integrated temperature sensor
- Robust and flexible wireless IDS pH measurement
- Securely measured values via signal processing in the sensor
- Easy electrode replacement
- Sensor quality monitoring by QSC function
- Memory for 10 calibration records
Specifications
Specifications
| Material | Plastic |
| pH Range | 0 to 14 |
| For Use With (Equipment) | MultiLine™ IDS and InoLab™ Multi IDS Meters |
| Includes | Plug head for AS/IDS-x cables or wireless modules IDS WLM-S |
| Internal Reference | Gel |
| Membrane | Ball |
| Temperature (Metric) Range | 0°C to 80°C |
| Length (Metric) | 120 mm |
| Diameter (Metric) | 12 mm |