Learn More
WTW™ SenTix™ 61 pH Electrode
Product Code. 10464071
Change view
Click to view available options
Connector Types:
Fixed Cable
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
10464071 Fixed Cable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10464071 Supplier WTW™ Supplier No. 103640
  • ONLINE EXCLUSIVE PRICE
    202.00€
    186.79€ / Pack of 1
    Save 15.21€
    7% Off
View all promotions
In Stock
Standard electrodes with gel and precision electrodes with liquid gel electrolyte for pH measurements in lab applications and in the field.
  • SenTix™: Standard electrodes with gel and precision electrodes with liquid gel electrolyte for pH measurements in lab applications and in the field
  • Maintenance-free gel reference system across the entire product life
  • Quick repsonse time by good connection to the medium for electrodes with liquid filled reference system
  • Best results thanks to high-quality membrane glasses
  • pH electrodes with gel electrolyte are ideal for portable measurement but also for routine measurement in the lab
TRUSTED_SUSTAINABILITY
Specifications
Connector Types Fixed Cable
Junction Platinum
Material Glass Shaft, Platinum Diaphragm
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type SenTix™ pH Electrode
For Use With (Equipment) Vario pH meter
Includes DIN electrode plug, 1m cable
Model SenTix™ 61
Storage Requirements Immerse the combination electrode in reference electrolyte (KCl 3mol/L, Ag+ free), pH combination electrodes must not be stored dry or in deionized water
Temperature (Metric) Range 0°C to 100°C
Length (Metric) Cable 1 m
Length (Metric) Stem 120 mm