Learn More
WTW™ SenTix™ 60 pH Electrode
Description
pH electrodes with gel electrolyte are ideal for portable measurement but also for routine measurement in the lab. The electrodes do not require any maintenance besides occasional cleaning and are therefore especially economical. For demanding measurements in the lab, there are SenTix™ electrodes with 3 mol/l KCl liquid electrolyte, easy to clean glass shaft and platinum diagphragm. These electrodes can also be used in difficult samples. And if you need an electrode with liquid electrolyte for portable measuring: The SenTix™ 51/52 with plastic shaft, integrated temperature sensor and ceramic diaphragma masters nearly every measuring task, even in the field.
Specifications
Specifications
| Connector Types | DIN, BNC |
| Junction | Platinum |
| Material | Glass Shaft, Platinum Diaphragm |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 100°C |
| Type | SenTix™ pH Electrode |
| For Use With (Equipment) | Vario pH meter |
| Includes | DIN/BNC electrode plug |
| Model | SenTix™ 60 |
| Storage Requirements | Immerse the combination electrode in reference electrolyte (KCl 3mol/L, Ag+ free), pH combination electrodes must not be stored dry or in deionized water |
| Show More |