Learn More
Cytiva Sentino™ Metricel™ Black Membrane
Product Code. 17194591
Click to view available options
Pore Size:
0.45 μm
0.8 μm
Unit Size:
Pack of 200
Product Code. 17194591
Brand: Cytiva 68125
Filter dispense packs for Sentino dispenser are ideal for high throughput microbiology testing labs looking for more ergonomic, faster ways to handle individual membranes.
  • Filter dispensers are ideal for high throughput microbiology testing labs looking for more ergonomic, faster ways to handle individual membranes
  • Designed to aseptically dispense one disc of membrane at the touch of a button
  • Gamma irradiated and ready to use
  • For research use only
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material (Membrane) Modified PES
Pore Size 0.8 μm
Sterility Non-sterile
No. per Pack 1000/Pk., Dispenser Pack of 200
Diameter (Metric)Top 47 mm