Learn More
Thermo Scientific™ Sensors and Replacement Batteries for Thermo Scientific™ Ultra-Low Temperature Freezers
Product Code. 11359311 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Sensor
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11359311 Sensor Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11359311 Supplier Thermo Scientific™ Supplier No. 290199
  • ONLINE EXCLUSIVE PRICE
    680.00€
    570.48€ / Pack of 1
    Save 109.52€
    16% Off
 Restricted Item
Estimated Shipment: 11-03-2027
Use these Thermo Scientific™ Sensors and Replacement Batteries with our ultra-low temperature freezers.
TRUSTED_SUSTAINABILITY
Specifications
Description Optional PT100 sensor for connection to building automation systems (Factory installed)
Type Sensor
For Use With (Equipment) Revco™ ExF/DxF, Forma™ 900, HERAfreeze™ HFU-B, TSE/TSD Series
Defrost Automatic