Learn More
Beskrivning
- This soft, rich foam soap has a dermatologically tested formula, skin-friendly pH and allergy-friendly ECARF certification, meaning it moisturises the skin and is suitable for frequent use.
- Save staff time with effortless refilling - certified by the Swedish Rheumatism Association, tested according to the Design for All test, published in the Swedish Design Research Journal, 2011.
- Dispenser reduces consumption by up to 50% and the bottle collapses for 70% less waste volume. (Essity test using Tork Foam Soap vs Tork Liquid Soap in Elevation dispenser. Based on Essity test)
- Help secure good hand hygiene - the factory-sealed bottle with a new pump for every refill helps reduce the risk of contamination and protect the formulations all the way to the hands.
Specifikationer
Specifikationer
| Product Type | Hand Soap |
| Color | White |
| Form | Liquid |
| Packaging | Plastic Bottle |
| Quantity | 6/Pk. |
| Size | 1 L |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?