Learn More
Thermo Scientific™ Sensititre™ YeastOne™ YO9 AST Plate
Product Code. 17454203
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 100
Product Code. 17454203
Brand: Thermo Scientific™ YO9
Simplify yeast testing with this Research Use Only Plate of nine antifungals, including Amphotericin B and Micafungin.
Simplify yeast susceptibility testing with the Thermo Scientific™ Sensititre™ YeastOne™ YO9 AST Plate with nine antifungals, including Amphotericin B and Micafungin. The Sensititre™ YeastOne™ YO9 AST Plate is intended for research use only.
Colorimetric Thermo Scientific™ alamarBlue™ agent provides reliable, easy and consistent endpoint determination with visual read option for efficient workflow and accurate results.
Achieve reliable, consistent results for Candida species with the Sensititre YeastOne YO9 AST Plate.
  • Room-temperature storage eliminates inventory control concerns
  • Individual packaging allows laboratory to test one plate at a time with no waste
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
  • 24-hour incubation time enables timely results for clinicians
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
0.5 McFarland Standard H2O (Part No. T3339)
Antimicrobial Agents Various
Broth Type YeastOne Broth (Part No. Y3462)
Description Sensititre YeastOne Research Use Only Plate, w/ Anidulafungin and Micafungin
Antibiotic Code Various
Product Type Microbiology Media
Test Type Yeast Susceptibility
Incubation Conditions 35°C O2
Incubation Time 24-25 hr or 48-72 hr for Cryptococcus* (*For research use only. Not for use in diagnostic procedures.)
Isolates Per Plate 1
No. per Pack 10
Quantity 10/Pk.
Reading Options Manual
Transfer Volume 20µL
Volume Per Well 100 μL
Species Candida species