missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sensititre™ Urinary CMV1BURF Vet AST Plate

Product Code. 17217270
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 17217270

Brand: Thermo Scientific™ YCMV1BURF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Perform accurate AST of urinary isolates of veterinary origin with this dual isolate, cost-effective plate.

Perform accurate AST with a urinary isolate-specific formulary with the dual isolate Thermo Scientific™ Sensititre™ Urinary CMV1BURF AST Plate. The dual isolate plate allows two animal isolates per plate reducing overall cost per test.

Achieve reliable, consistent results for non-fastidious Gram-positive and Gram-negative isolates of veterinary origin with Sensititre Urinary CMV1BURF AST Plate.

  • Extended shelf life, room temperature storage eliminates inventory control concerns.
  • Individual packaging allows laboratory to test one plate at a time with no waste.
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology.
  • Cost-effective: the dual isolate plate allows two patient samples per plate reducing the cost per test.
  • Automated or manual read options are available for ultimate flexibility.

Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents Various
Description Sensititre Urinary MIC Plate
Antibiotic Code Various
Product Type AST Plate
Target Organism Class Non-fastidious, Gram-positive and Gram-negative Veterinary Isolates
Test Type Veterinary AST, Minimum Inhibitory Concentration (MIC)
Isolates Per Plate 2
Quantity 10/Pk.
Reading Options Autoread or manual
Specimen Type Animal Urinary Isolates, Two Isolates per Plate
Species All
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.