Learn More
Sensititre™ Gram Positive EUSTAPF AST Plate
Product Code. 17731989
Click to view available options
Broth Type:
MHB (part no. T3462)
Unit Size:
Pack of 10
Product Code. 17731989
Brand: Thermo Scientific Oxoid YEUSTAPF
Perform susceptibility testing of non-fastidious Staphylococcus species on a single, convenient plate which meets EUCAST breakpoints
Perform susceptibility testing of non-fastidious Staphylococcus species isolates on a single platform with Thermo Scientific™ Sensititre™ Gram-Positive EUSTAPF AST Plate. The convenient standard AST plate meets EUCAST breakpoints for relevant antimicrobials on the plate.
Achieve reliable, consistent antimicrobial susceptibility results for non-fastidious Staphylococcus species isolates with the Sensititre Gram-Positive EUSTAPF AST Plate. Generate clinically relevant data, detect emerging resistance, and help guide timely and accurate therapy intervention by utilizing micro-broth dilution.
  • Monitor susceptibility among Gram-negative isolates with full-range, accurate MIC values
  • For ultimate flexibility automated or manual read options are available for ultimate flexibility
  • Eliminate inventory control concerns with extended shelf life (18 to 24 months) and room temperature storage.
The Sensititre System is a leader in AST solutions, offering flexible, customizable testing options for laboratories of all sizes.
  • Scalable: flexible instrumentation options designed to be modular in order to meet the unique needs of individual lab space, workload, and budget requirements
  • Flexible: over 240 antimicrobials available in extended dilution ranges, access to the newest antimicrobials, and the ability to consolidate AST onto one platform
  • Tailor-made: design your own AST panel to meet specific formulary requirements and prescription protocols and to clinicians that are based on real-time trends and local epidemiology
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
0.5 McFarland Standard Demineralized Water (part no. T3339)
Antimicrobial Agents Various
Broth Type MHB (part no. T3462)
Description Sensititre Gram Positive EUSTAPF AST Plate
Antibiotic Code Various
Product Type AST Plate
Test Type Minimum Inhibitory Concentration (MIC)
Incubation Conditions 35˚C; O2
Incubation Time 18-24 hrs
Isolates Per Plate 1
Packaging Type Box
Quantity 10/Box
Reading Options Autoread or manual
Transfer Volume 10 µL
Volume Per Well 50 μL
Species Non-fastidious Staphylococcus