Learn More
Sensititre™ Gram Positive EUSTAPF AST Plate
Description
Perform susceptibility testing of non-fastidious Staphylococcus species on a single, convenient plate which meets EUCAST breakpoints
Perform susceptibility testing of non-fastidious Staphylococcus species isolates on a single platform with Thermo Scientific™ Sensititre™ Gram-Positive EUSTAPF AST Plate. The convenient standard AST plate meets EUCAST breakpoints for relevant antimicrobials on the plate.
Achieve reliable, consistent antimicrobial susceptibility results for non-fastidious Staphylococcus species isolates with the Sensititre Gram-Positive EUSTAPF AST Plate. Generate clinically relevant data, detect emerging resistance, and help guide timely and accurate therapy intervention by utilizing micro-broth dilution.
- Monitor susceptibility among Gram-negative isolates with full-range, accurate MIC values
- For ultimate flexibility automated or manual read options are available for ultimate flexibility
- Eliminate inventory control concerns with extended shelf life (18 to 24 months) and room temperature storage.
The Sensititre System is a leader in AST solutions, offering flexible, customizable testing options for laboratories of all sizes.
- Scalable: flexible instrumentation options designed to be modular in order to meet the unique needs of individual lab space, workload, and budget requirements
- Flexible: over 240 antimicrobials available in extended dilution ranges, access to the newest antimicrobials, and the ability to consolidate AST onto one platform
- Tailor-made: design your own AST panel to meet specific formulary requirements and prescription protocols and to clinicians that are based on real-time trends and local epidemiology
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| 0.5 McFarland Standard | Demineralized Water (part no. T3339) |
| Antimicrobial Agents | Various |
| Broth Type | MHB (part no. T3462) |
| Description | Sensititre Gram Positive EUSTAPF AST Plate |
| Antibiotic Code | Various |
| Product Type | AST Plate |
| Test Type | Minimum Inhibitory Concentration (MIC) |
| Incubation Conditions | 35˚C; O2 |
| Incubation Time | 18-24 hrs |
| Isolates Per Plate | 1 |
| Show More |