Learn More
Sensititre™ Gram Negative GNX2F AST Plate
Shop All Thermo Scientific ProductsDescription
Perform susceptibility testing of Gram-negative isolates on a single platform including colistin and Polymixin B with this research use only plate.
Perform susceptibility testing of non-fastidious Gram-negative isolates on a single platform with Thermo Scientific™ Sensititre™ Gram Negative GNX2F AST Plate, designed for research use only.
Decrease costs associated with multiple test methods with 21 antimicrobial compounds on a single plate, including colistin and polymixin B.
Test most bacteria, including challenging resistant organisms, on a single platform with Sensititre Gram Negative GNX2F AST Plate.
- Automated or manual read options are available for ultimate flexibility
- Extended shelf life and room-temperature storage simplify inventory control
- Individual packaging allows laboratory to test one plate at a time with no waste
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| 0.5 McFarland Standard | H2O (Part No. T3339) |
| Antimicrobial Agents | Various |
| AST Type | MIC |
| Broth Type | MHB (Part No. T3462) |
| Description | Sensititre Gram Negative Research Use Only MIC Plate for CRE Testing |
| Antibiotic Code | Various |
| Product Type | AST Plate |
| Target Organism Class | Non-fastidious, Gram-negative Organisms |
| Test Type | Minimum Inhibitory Concentration (MIC), Carbapenem-resistant Enterobacterales (CRE) |
| Incubation Conditions | 35°C O2 |
| Show More |