Learn More
Thermo Scientific™ Sensititre™ Gram Negative MIC Plate
Product Code. 15334869
Click to view available options
Description:
MIC Plate
Unit Size:
Pack of 10
Product Code. 15334869
Brand: Thermo Scientific™ YGN2F
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Utilize the latest antimicrobials, including Doripenem and Tigecycline, for accurate, economical results with Thermo Scientific™ Sensititre™ Gram Negative MIC Plates.
Utilize the latest antimicrobials, including Doripenem and Tigecycline, for accurate, economical results with Sensititre Gram Negative MIC Plates.
  • Multi-isolate format decreases cost per test
  • Automated, semiautomated or manual read options are available for ultimate flexibility
  • Extended shelf life and room-temperature storage simplify inventory control
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
0.5 McFarland Standard H2O (Part No. T3339)
Antimicrobial Agents Various
Broth Type MHB (Part No. T3462)
Description MIC Plate
Antibiotic Code Various
Product Type MIC Plate
Target Organism Class Gram-negative Bacteria
Test Type Minimum Inhibitory Concentration (MIC), Breakpoint
For Use With (Application) Gram negative organisms: Enterobacteriaceae, Non-Enterobacteriaceae, Proteus sp.
Incubation Conditions 35°C O2
Incubation Time 18 hr
Isolates Per Plate 1
No. per Pack 10
Packaging Type Box
Quantity 10/Pk.
Reading Options Autoread or manual
Transfer Volume For Enterobacteriaceae and Non-Enterobacteriaceae: 10μL, 30μL* (*Aid in detection of resistance mechanisms for gram negative organisms; see pack insert). For Proteus sp.: 1μL
Volume Per Well 50 μL
Species Multiple