Learn More
Thermo Scientific™ Sensititre™ Gram Negative EUMDROXF AST Plate
Product Code. 17805425 Shop All Thermo Scientific Products
Product Code. 17805425
Brand: Thermo Scientific™ EUMDROXF
Confidently perform AST of Gram-negative isolates on a single plate containing cefiderocol,eravacycline, imipenem/relebactam and meropenem/vaborbactam.
Confidently perform susceptibility testing of multi-drug resistant Gram-negative isolates on a single plate which includes cefiderocol, eravacycline, imipenem/relebactam and meropenem/vaborbactam using Thermo Scientific™ Sensititre™ Gram Negative EUMDROXF AST Plates, designed for in-vitro diagnostic use.
This broth microdilution plate provides additional treatment options where traditional antibiotics are no longer effective. Cefiderocol, eravacycline imipenem/relebactam meropenem/vaborbactam represent four of the latest treatment options for hard to treat infections like complicated urinary tract infections (cUTIs), including pyelonephritis.
Expand your Gram-negative organism susceptibility testing options by accessing the latest antibiotics including cefiderocol, eravacycline, imipenem/relebactam and meropenem/vaborbactam
The Gram Negative EUMDROXF AST Plate delivers results directly from broth microdilution (BMD), the gold standard1 phenotypic method for antimicrobial Succeptibility Testing (AST).
·        New plate format : including Cefiderocol, Eravacycline, Imipenem/Relebactam and Meropenem/Vaborbactam
·        Efficient: BMD provides accurate MIC results guiding treatment optimization, reducing unnecessary testing and protocols
·        Accurate: Meets the latest EUCAST breakpoints
·        Quantitative: Sensititre MIC is determined using direct measurement rather than relying on extrapolated data, thereby providing an accurate MIC result, and not an estimate.
·        Flexible:  All contained antibiotics are also available in customizable plate options, along with access to over 300 antimicrobials, to enable meeting local formulary and Stewardship program requirements
Thermo Fisher Scientific is a leader in providing AST solutions that help in the fight against antimicrobial resistance, offering high performance automated and manual test options and one of the largest, most up-to-date selections of antimicrobials for susceptibility testing.
The Sensititre System utilizes gold standard-level MIC accuracy, providing superior quality and reproducibility for accurate results the first time.  Broth microdilution is the reference method1 for AST, and in many cases2 the most reliable phenotypic method to guide therapeutic decisions.
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Various
Description Sensititre Gram Negative EUMDROXF AST Plate
Antibiotic Code Various
Product Type AST Plate
Target Organism Class Multidrug-resistant, Gram-negative Organisms
Test Type Minimum Inhibitory Concentration (MIC)
Incubation Time 18 – 24 hrs
Packaging Type Box
Quantity 10/Box
Reading Options Manual and autoread