Learn More
Thermo Scientific™ Sensaphone™ Telephone Dialing Systems
Product Code. 12773838 Shop All Thermo Scientific Products
Click to view available options
Type:
Four Channels
Electrical Requirements:
220 V 50 Hz
Unit Size:
Each
Not approved for sales requiring CE mark.
Product Code. 12773838
Brand: Thermo Scientific™ 400182
Not approved for sales requiring CE mark.
Remotely monitor equipment from any standard touch-tone phone with this telephone dialing system will report the status of every monitored condition.
Call the lab or office from anywhere and Thermo Scientific™ Sensaphone™ Telephone Dialing Systems will state the status of every monitored condition. These systems interface with any standard touch-tone phone to notify of alarm conditions.
  • Can be used with Model 1535 Monitor/Alarm System that features programmable audible/visual alarms and updates/displays equipment conditions on an LCD readout
  • Battery back-up is standard
  • Battery life up to 15 hr. after AC power has failed; up to 18 months with AC power on
Requires:
Six D-size alkaline batteries
TRUSTED_SUSTAINABILITY
Specifications
Description Four pre-programmed numbers
Type Four Channels
For Use With (Equipment) Model 1535 Monitor/Alarm System
Monitoring Options Alarm
Voltage 220 V
Electrical Requirements 220 V 50 Hz
Frequency 50 Hz