Learn More
Semperit™ Sempermed™ Synthetic Polyisoprene Syntegra IR Surgical Gloves
Description
- Made from Synthetic Polyisopren
- Powder-free
- Free from natural rubber latex
- Free from allergenic proteins - this removes the hazard of sensitisation right from the start, especially for
atopic persons. - Quick and easy to put on in every situation due to its elastic material.
- Lining includes a special net-like structure that reduces friction resistance when donning the gloves
- Guarantee the easy donning of two pairs of gloves, without any loss of tactile properties.
- Fully anatomical shape with curved fingers corresponds with the natural, relaxed hand posture
- Micro-roughened surface offers optimum grip for perfect instrument handling, even in a wet environment
Specifications
Specifications
| Certifications/Compliance | EN 1041, EN 980, EN 455/3, ISO 15223, EN 455 Class 1 |
| Color | Cream |
| Disposable | Single-use |
| Latex Free | Yes |
| Material | Synthetic Polyisoprene |
| Powdered | Powdered |
| Size | 8.5 |
| Sterility | Sterile |
| Thickness (Metric) | 0.19 to 0.24 mm |
| For Use With (Application) | For perfect instrument handling, even in a moist environment, fully anatomical fit guarantee a relaxed hand position for less tiring work |
| Show More |