Learn More
Brady™ Self-laminating Vinyl Wrap Around Labels
Mark cables and wires with the B-427 self-laminating vinyl label material, designed with a clear self-laminating protective layer. Print almost anywhere with a durable Brady™ portable label printer.
30.23€ - 91.42€
Specifications
Certifications/Compliance | UL Recognized to UL 2238, RoHS Compliant to 2011/65/EU (Pass 2015/863) |
---|---|
Adhesive Type | Permanent Acrylic |
Finish | Matte |
Material | Self-laminating Vinyl |
Product Type | Labels |
Product Code | Brand | Color | Dimensions (LxW) | For Use With (Equipment) | Format | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Dimensions (LxW) | For Use With (Equipment) | Format | Price | Quantity & Availability | ||
17930680
|
Brady™
170748 |
Black on White | 1.1 x 0.25 in. | BMP41, BMP51, M511 | Die-cut |
32.94€
Pack of 240 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17990760
|
Brady™
170742 |
Black on White/Transparent | 0.75 x 0.5 in. | BMP41, BMP51, M511 | Die-cut |
48.48€
Pack of 360 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17970720
|
Brady™
170751 |
Black on White/Transparent | 0.75 x 1 in. | BMP41, BMP51, M511 | Die-cut |
79.85€
Pack of 340 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17980710
|
Brady™
170755 |
Black on White/Transparent | 0.5 x 1 in. | BMP41, BMP51, M511 | Die-cut |
45.05€
Pack of 360 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17930720
|
Brady™
170752 |
Black on White/Transparent | 1 x 0.75 in. | BMP41, BMP51, M511 | Die-cut |
38.52€
Pack of 200 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17940720
|
Brady™
170750 |
Black on White/Transparent | 1 x 1 in. | BMP41, BMP51, M511 | Die-cut |
62.08€
Pack of 260 |
In Stock
Log in to see stock. |
||
17841769
|
Brady™
170869 |
Black on White/Transparent | 1 x 1.75 in. | BMP51, M511 | Die-cut |
50.86€
Pack of 140 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17900710
|
Brady™
170741 |
Black on White/Transparent | 1.25 x 0.5 in. | BMP41, BMP51, M511 | Die-cut |
36.97€
Pack of 200 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17920720
|
Brady™
170744 |
Black on White/Transparent | 1.25 x 1 in. | BMP41, BMP51, M511 | Die-cut |
53.89€
Pack of 200 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17801789
|
Brady™
170864 |
Black on White/Transparent | 1.25 x 1.5 in. | BMP51, M511 | Die-cut |
61.60€
Pack of 200 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17930710
|
Brady™
170747 |
Black on White/Transparent | 1.5 x 0.5 in. | BMP41, BMP51, M511 | Die-cut |
30.23€
Pack of 180 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17900720
|
Brady™
170745 |
Black on White/Transparent | 1.5 x 0.75 in. | BMP41, BMP51, M511 | Die-cut |
45.06€
Pack of 180 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17970670
|
Brady™
170743 |
Black on White/Transparent | 1.5 x 1 in. | BMP41, BMP51, M511 | Die-cut |
51.98€
Pack of 180 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17861789
|
Brady™
170866 |
Black on White/Transparent | 1.5 x 1.5 in. | BMP51, M511 | Die-cut |
64.11€
Pack of 180 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17841789
|
Brady™
170870 |
Black on White/Transparent | 1.5 x 1.75 in. | BMP51, M511 | Die-cut |
48.00€
Pack of 140 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17950720
|
Brady™
170749 |
Black on White/Transparent | 2.5 x 1 in. | BMP41, BMP51, M511 | Die-cut |
50.81€
Pack of 110 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||
17841759
|
Brady™
170865 |
Black on White/Transparent | 2.5 x 1.5 in. | BMP51, M511 | Die-cut |
49.28€
Pack of 80 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17990710
|
Brady™
170754 |
Black on White/Transparent | 4 x 1 in. | BMP41, BMP51, M511 | Die-cut |
48.95€
Pack of 70 |
Estimated Shipment: 09-07-2024
Log in to see stock. |
||
17851719
|
Brady™
170867 |
Black on White/Transparent | 4 x 1.5 in. | BMP51, M511 | Die-cut |
58.78€
Pack of 70 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17811719
|
Brady™
170868 |
Black on White/Transparent | 6 x 1.5 in. | BMP51, M511 | Die-cut |
48.50€
Pack of 40 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17901950
|
Brady™
170740 |
Black on White/Transparent | 25 ft. x 0.75 in. | BMP41, BMP51, M511 | Continuous |
72.27€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17811729
|
Brady™
170861 |
Black on White/Transparent | 25 ft. x 1 in. | BMP51, M511 | Continuous |
87.60€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17881799
|
Brady™
170862 |
Black on White/Transparent | 25 ft. x 1.25 in. | BMP51, M511 | Continuous |
89.14€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17811779
|
Brady™
170863 |
Black on White/Transparent | 25 ft. x 1.5 in. | BMP51, M511 | Continuous |
91.42€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17910720
|
Brady™
170746 |
Black on Yellow, Clear | 1.5 x 1 in. | BMP41, BMP51, M511 | Die-cut |
51.98€
Pack of 180 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
17920680
|
Brady™
170753 |
Black on Yellow, Clear | 2.5 x 1 in. | BMP41, BMP51, M511 | Die-cut |
50.81€
Pack of 110 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
17970710
|
Brady™
170756 |
Black on Yellow, Clear | 4 x 1 in. | BMP41, BMP51, M511 | Die-cut |
48.95€
Pack of 70 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||
Description
- Designed for tightly-curved surfaces such as wires and cables
- Self-extinguishing and resistant to abrasion, solvents, oil, grease, weather and extreme temperatures
- Made with a permanent acrylic adhesive that works great on smooth, rough, powder-coated and irregular surfaces
- Made with a clear, non-printable overlaminate for increased protection
Specifications
UL Recognized to UL 2238, RoHS Compliant to 2011/65/EU (Pass 2015/863) | |
Matte | |
Labels | |
Data and Telecommunications Labeling, Electrical Labeling, Laboratory Labeling, Wire and Cable Labeling | |
0.004 in. |
Permanent Acrylic | |
Self-laminating Vinyl | |
Non-sterile | |
-94°F to +158°F |