Learn More
Brady™ Self-laminating Polyester Labels for M710
Product Code. 17848064
Click to view available options
Dimensions (LxW):
15 x 40 mm
22.86 x 22.86 mm
25.4 x 25.4 mm
38.1 x 15.24 mm
41.28 x 15.24 mm
44.45 x 25.4 mm
45.72 x 9.53 mm
48.26 x 15.24 mm
66.68 x 15.24 mm
66.68 x 25.4 mm
Color:
White
White, Transparent
Shape:
Rectangle
Rectangle with Circle
Square
Unit Size:
Pack of 100
Pack of 150
Pack of 250
Pack of 500
Product Code. 17848064
Brand: Brady™ 311977
Printer Labels for use with Brady™ Printer M710.
  • Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance and high/low temperature performance
Specifications
Certifications/Compliance ASTM D 1000
Adhesive Type Permanent Acrylic
Coating None
Color White, Transparent
Finish Matte
Format Label
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Self-laminating Tag
Sterility Non-sterile
For Use With (Application) Laboratory Labeling
For Use With (Equipment) M710, BMP71
Labels per Row 1 Label
Min. Operating Temperature (Metric) -196°C
Quantity 150 Labels
Shape Rectangle with Circle
Temperature Range (English) -321°F to +230°F
Temperature Range (Metric) -196°C to +110°C
Thickness (English) 0.0019 in.
Thickness (Metric) 0.048 mm
Dimensions (LxW) 41.28 x 15.24 mm
Length (English) 1.625 in.
Width (English) 0.6 in.
Width (Metric) 15.24 mm
Width (English) Printable Area 15.24 mm
Width (Metric) Printable Area 15.24 mm
Height (English) Printable Area 1.18 in.
Height (Metric) Printable Area 30 mm
Diameter (English) 0.5 in.
Diameter (Metric) 12.7 mm
Alternate Part Number M7-116-461