Learn More
Brady™ Self-laminating Polyester Labels for M710
Product Code. 17827884
Click to view available options
Dimensions (LxW):
15 x 40 mm
22.86 x 22.86 mm
25.4 x 25.4 mm
38.1 x 15.24 mm
41.28 x 15.24 mm
44.45 x 25.4 mm
45.72 x 9.53 mm
48.26 x 15.24 mm
66.68 x 15.24 mm
66.68 x 25.4 mm
Color:
White
White, Transparent
Shape:
Rectangle
Rectangle with Circle
Square
Unit Size:
Pack of 100
Pack of 150
Pack of 250
Pack of 500
Product Code. 17827884
Brand: Brady™ 311975
Only null left
Add to basket
Add to basket
Printer Labels for use with Brady™ Printer M710.
  • Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance and high/low temperature performance
Specifications
Certifications/Compliance ASTM D 1000
Adhesive Type Permanent Acrylic
Coating None
Color White, Transparent
Finish Matte
Format Label
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Self-laminating Tag
Sterility Non-sterile
For Use With (Application) Laboratory Labeling
For Use With (Equipment) M710, BMP71
Labels per Row 1 Label
Min. Operating Temperature (Metric) -196°C
Quantity 250 Labels
Shape Rectangle
Temperature Range (English) -321°F to +230°F
Temperature Range (Metric) -196°C to +110°C
Thickness (English) 0.0019 in.
Thickness (Metric) 0.048 mm
Dimensions (LxW) 44.45 x 25.4 mm
Length (English) 1.75 in.
Width (English) 1 in.
Width (Metric) 25.4 mm
Width (English) Printable Area 25.4 mm
Width (Metric) Printable Area 25.4 mm
Height (English) Printable Area 1 in.
Height (Metric) Printable Area 25.4 mm
Alternate Part Number M7-106-461