Learn More
Brady™ Self-laminating Polyester Labels for M710
Product Code. 17808034
Click to view available options
Dimensions (LxW):
15 x 40 mm
22.86 x 22.86 mm
25.4 x 25.4 mm
38.1 x 15.24 mm
41.28 x 15.24 mm
44.45 x 25.4 mm
45.72 x 9.53 mm
48.26 x 15.24 mm
66.68 x 15.24 mm
66.68 x 25.4 mm
Color:
White
White, Transparent
Shape:
Rectangle
Rectangle with Circle
Square
Unit Size:
Pack of 100
Pack of 150
Pack of 250
Pack of 500
Product Code. 17808034
Brand: Brady™ 311983
Printer Labels for use with Brady™ Printer M710.
  • Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance and high/low temperature performance
Specifications
Certifications/Compliance ASTM D 1000
Adhesive Type Permanent Acrylic
Coating None
Color White, Transparent
Finish Matte
Format Label
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Self-laminating Tag
Sterility Non-sterile
For Use With (Application) Laboratory Labeling
For Use With (Equipment) M710, BMP71
Labels per Row 1 Label
Min. Operating Temperature (Metric) -196°C
Quantity 250 Labels
Shape Square
Temperature Range (English) -321°F to +230°F
Temperature Range (Metric) -196°C to +110°C
Thickness (English) 0.0019 in.
Thickness (Metric) 0.048 mm
Dimensions (LxW) 25.4 x 25.4 mm
Length (English) 1 in.
Width (English) 1 in.
Width (Metric) 25.4 mm
Width (English) Printable Area 25.4 mm
Width (Metric) Printable Area 25.4 mm
Height (English) Printable Area 0.38 in.
Height (Metric) Printable Area 9.53 mm
Alternate Part Number M7-19-461