Learn More
Testo™ Self-Adhesive Reflective Tape
Product Code. 11301254
Change view
Click to view available options
Length (Metric):
150 mm
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11301254 150 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11301254 Supplier Testo™ Supplier No. 0554 0493
  • ONLINE EXCLUSIVE PRICE
    69.30€
    28.00€ / Pack of 5
    Save 41.30€
    59% Off
Estimated Shipment: 18-11-2026
Use this reflective tape for a variety of applications. Testo™ Self-Adhesive Reflective Tape is designed for use with Testo tachometers.
  • Self-adhesive reflectors
  • 150mm long
  • Available in packs of 5
Specifications
For Use With (Equipment) For tachometers
Length (Metric) 150 mm