Learn More
Witeg™ Security Washing Bottle
Product Code. 10363542
Change view
Click to view available options
Capacity (Metric):
500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10363542 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10363542 Supplier Witeg™ Supplier No. 2 418 250
  • ONLINE EXCLUSIVE PRICE
    342.82€
    308.53€ / Pack of 1
    Save 34.29€
    10% Off
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
For the rinsing of various items in the laboratory.
  • With screw-threads
  • Porosity of P0
  • With two SVS-tubing connections, GL 14, center neck GL 32
  • With septum
  • Made of borosilicate glass
  • Washing intensity of 130 L/h
Specifications
Capacity (Metric) 500 mL