Learn More
3M™ SecureFit™ X5000 Safety Helmet, Vented, Reflective, CE
Product Code. 17345323
Click to view available options
Color:
Black
Blue
Green
Hi-vis Green
Orange
Red
White
Yellow
Unit Size:
Pack of 4
Product Code. 17345323
Brand: 3M™ 7100175100
Only null left
Add to basket
Add to basket
3M™ SecureFit™ Safety Helmet has six different settings to adjust position and a 4-point chin strap selector system. Vented and brimless safety helmet with 3M™ UVicator™ Sensor that indicates damaging levels of UV light.
  • Secure, comfortable head protection for working at heights and other hazardous industrial jobs
  • Suspension system incorporates exclusive patented 3M™ Pressure Diffusion Technology
  • 4-point chin strap selector system unique to 3M allows wearer to switch between EN 12492 (helmets for mountaineers) and EN 397 (industrial safety helmets) standards
  • 6-point ratchet suspension allows wearer to customise height and fit with multiple levels of vertical adjustment
  • Magnetic buckle clip as standard
  • Inspired by climbing helmets with a brimless design to give you a clear upward view
  • Accessory slots and clips are compatible with a variety of 3M face shields, earmuffs and other accessories
  • 3M™ Uvicator™ sensor disc tells you when your hard hat has been over exposed to UV-radiation and it's time to replace it
  • Certified to either EN12492 or EN397 (depending on chin strap configuration)
  • Approved for lateral deformation (LD)
  • UV-stabilized ABS
TRUSTED_SUSTAINABILITY
Specifications
Color Hi-vis Green
For Use With (Application) Construction, Forestry, Oil, and Gas
Material ABS
Product Type Safety Helmet
Suspension Type Pressure Diffusion 6 Point Ratchet
Ventilated Ventilated