Learn More
3M™ SecureFit™ Safety Glasses, Anti-Fog / Anti-Scratch Coating (K&N)
missing translation for 'orderingAttributeHoverText'
Frame Color:
Black/Yellow
Brown
Gray/Blue
Grey
Red/Gray
Lens Tint:
Amber
Brown
Clear
Gray
Indoor/Outdoor Gray
Orange
Description:
SF401SGAF-BLU-EU
SF401SGAF-BLU-F-EU, Includes removable foam-lined gasket
SF401SGAF-RED-EU
SF402SGAF-BLU-EU
SF403SGAF-YEL-EU
SF405SGAF-BLA-EU
SF406SGAF-BLA-EU
SF407SGAF-BLU-EU
SF601SGAF/FI-EU
missing translation for 'unitSize'
Each
Pack of 20
Beschreibung
- Diffuse pressure over the ear with 3M™ Pressure Diffusion Temple Technology
- Soft, adjustable nose pads provide added comfort
- Flat and flexible temples combine easily with earmuffs and provide solid side protection
- Wraparound design gives the user a wide field of vision
- 3M™ Scotchgard™ Anti-Fog Coating offers superior anti-fog and anti-scratch properties
- Excellent protection against Ultraviolet (UV) radiation
- 19 g lightweight design
Spezifikation
Spezifikation
| Lens Coating | Anti-fog |
| Description | SF401SGAF-RED-EU |
| Lens Tint | Clear |
| Frame Color | Red/Gray |
| Includes | |
| Quantity | 20/Cs. |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur