missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Secure-it™ Tamper-Proof Labels

Product Code. 11981431 Shop All Bürkle Products
Change view
Click to view available options
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
11981431 Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11981431 Supplier Buerkle™ Supplier No. 53035010

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Produces a silver checkerboard pattern on both the subsurface and surface of the sticker itself when the seal has been broken. Buerkle™ Secure-it™ Tamper Proof Labels act as originality seals and protects samples from interference during storage or transport, for sealing containers with critical or dangerous reagents or chemicals in the lab, as well as for sealing documentation, shipments and so much more.

Safe-it and Secure-it safety labels are used as originality seals and as protection from interference with samples during storage or transport, for sealing containers with critical or dangerous reagents or chemicals in the lab, as well as for sealing documentation, shipments and so much more. When the Safe-it security seal has been broken, a silver checkerboard pattern appears on both the subsurface and surface of the sticker itself.
  • With labeling field
  • With stop sign pattern
  • Visible proof of tampering
  • Size 95 x 20mm
  • 500 labels per roll
TRUSTED_SUSTAINABILITY

Specifications

Description Produces a bright purple stop sign when seal is broken
Color Purple
For Use With (Application) Protecting samples from interference
Quantity 500/Roll
Width (Metric) 95 mm
Height (Metric) 20 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.