Learn More
3M™ Secure Click™ Reusable Full Face Respirator
Product Code. 17976640 Shop All 3M Products
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17976640 Small Each
17956640 Medium Each
17966640 Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17976640 Supplier 3M™ Supplier No. 7100262627
  • ONLINE EXCLUSIVE PRICE
    487.00€
    327.06€ / Pack of 1
    Save 159.94€
    32% Off
Estimated Shipment: 09-12-2026
The Secure Click™ Reusable Full Face Mask FF-800 Series helps protect against particulates and a wide variety of gases and vapors when used with approved Secure Click™ filters.
  • The quick, easy, one-touch positive pressure push button seal check helps provide improved confidence in respirator fit
  • Two dual-flow gas and vapor filters on each respirator combine to create the world's first quad flow gas and vapor filter system with four airflow paths providing easy breathing and comfort
  • Large lens for excellent field of vision, and treated with Scotchgard™ Protector for improved paint and stain resistance
  • Six-strap harness helps provide a secure comfortable fit
  • Speaking diaphragm is designed to help provide easy communication
  • Recommended applications include assembly, chemical clean-up, chemical handling, chipping, chiseling, cleaning, furnace operations, grinding, machining, masonry, painting, pouring/casting, sanding and sawing.
  • Available in three sizes
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Filter Type Secure Click™ Filters
Size Small