missing translation for 'onlineSavingsMsg'
Learn More

sec-Butyl methyl ether, 99%

Product Code. 15460228
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Unit Size:
100mL
25mL
5mL
This item is not returnable. View return policy

Product Code. 15460228

Brand: Thermo Scientific Alfa Aesar H53427.AC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6795-87-5 | C5H12O | 88.15 g/mol

sec-Butyl methyl ether is used as a solvent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
sec-Butyl methyl ether is used as a solvent in organic synthesis.

Solubility
Miscible with water, alcohol, ether and acetone.

Notes
Incompatible with strong oxidizing agents. Avoid conditions like flames, sparks and direct sunlight.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6795-87-5
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00190205
InChI Key FVNIMHIOIXPIQT-UHFFFAOYNA-N
Synonym sec-butyl methyl ether, butane, 2-methoxy, methyl sec-butyl ether, ether, sec-butyl methyl, 2-butyl methyl ether, methyl 1-methylpropyl ether, s-butyl methyl ether, butane, 2-methoxy-9ci, butane,2-methoxy, acmc-1b64a
PubChem CID 23238
IUPAC Name 2-methoxybutane
SMILES CCC(C)OC

Specifications

Density 0.742
Boiling Point 57°C
Flash Point −30°C (−22°F)
Refractive Index 1.372
Quantity 25 mL
UN Number UN2350
Solubility Information Miscible with water,alcohol,ether and acetone.
Formula Weight 88.15
Percent Purity 99%
Chemical Name or Material sec-Butyl methyl ether

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado