missing translation for 'onlineSavingsMsg'
Learn More

sec-Butyl Mercaptan, TRC

Product Code. 30336351
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336351 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336351 Supplier Toronto Research Chemicals Supplier No. TRCB6926901G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material sec-Butyl Mercaptan
CAS 513-53-1
Molecular Formula C4H10S
Quantity 1 g
Synonym (±)-2-Butanethiol, 1-Methyl-1-propanethiol, 2-Butyl mercaptan, 2-Butylthiol, 2-Mercaptobutane, 3-Methyl-2-propanethiol, NSC 78417, s-Butyl mercaptan, sec-Butanethiol, sec-Butyl mercaptan, sec-Butyl thioalcohol, sec-Butylthiol, 2-Butanethiol, (±)-2-Butanethiol, 1-Methyl-1-propanethiol, 2-Butyl mercaptan, 2-Butylthiol, 2-Mercaptobutane, 3-Methyl-2-propanethiol, NSC 78417, s-Butyl mercaptan, sec-Butanethiol, sec-Butyl thioalcohol, sec-Butylthiol, 2-Butanethiol
InChI Formula InChI=1S/C4H10S/c1-3-4(2)5/h4-5H,3H2,1-2H3
SMILES CCC(C)S
IUPAC Name butane-2-thiol
Molecular Weight (g/mol) 90.19
Formula Weight 90.0503
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.