Learn More
Buerkle™ Sealing Sleeves for Buerkle™ Zone Samplers
Product Code. p-7230565 Shop All Bürkle Products
Click to view available options
Diameter (Metric) Outer Top:
25 mm
50 mm
Unit Size:
1 set
Product Code. 10770763
Brand: Buerkle™ 53170011
Sealing sleeves made of PP, sterilisable, they seal individual chambers when filling, emptying or transporting. The sealing sleeves are easy to slide on and they automatically clip into position.
Sealing sleeves made of PP, sterilizable, which seal individual chambers when filling, emptying or transporting. The sealing sleeves are easy to slide on and automatically clip into position. Length 150 mm
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
For Use With (Equipment) Zone Sampler
Diameter (Metric) Outer Top 25 mm
Length (Metric) 150 mm