Learn More
Prestige Medical™ Sealing Gasket
Product Code. 13466429
Change view
Click to view available options
Quantity:
Pk of 4
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13466429 Pk of 4 Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13466429 Supplier Prestige Medical™ Supplier No. 219748
  • ONLINE EXCLUSIVE PRICE
    195.69€
    164.19€ / Pack of 4
    Save 31.50€
    16% Off
Estimated Shipment: 14-12-2026
Sealing gaskets For Classic models with a sterlising temperature of 121°C/126°C, green.
  • An important safety feature of your autoclave
  • Correct maintenance and replacement of your sealing gasket will help prevent unnecessary breakdowns and expensive down time
Specifications
Quantity Pk of 4
Type Sealing Gasket