Learn More
Cytiva SE260/SE250 MiniVertical Electrophoresis System Replacement Parts: Spineless Combs
Product Code. 10783657 Shop All Cytiva Products
Click to view available options
No. of Wells:
5
10
15
Thickness (Metric):
0.07 cm
0.1 cm
0.15 cm
Volume (Metric) Well:
19.1μL/ mm
2.9μL/ mm
4.4μL/ mm
4.8μL/ mm
7.2μL/ mm
9.5μL/ mm
Width (Metric):
0.29 cm
0.48 cm
1.3 cm
Unit Size:
Each
Product Code. 10783657
Brand: Cytiva SE211A151.0
Only null left
Specifications
For Use With (Equipment) SE 250, SE 260 Mini Format Vertical Gel Electrophoresis Unit
Type Spineless Comb
Material PTFE
No. of Wells 15
Thickness (English) 0.03 in.
Thickness (Metric) 0.1 cm
Volume (Metric) Well 2.9μL/ mm
Width (Metric) 0.29 cm
Depth (English) 0.03 in.
Depth (Metric) 0.1 cm