missing translation for 'onlineSavingsMsg'
Learn More

Melford SDS, Powder

Product Code. p-200087055
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
5 kg
25 kg
Unit Size:
100g
1kg
250g
25kg
500g
5kg
This item is not returnable. View return policy

Product Code. 30018375

Brand: Melford L22010250.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ionic detergent for denaturing proteins prior to gel electrophoresis.

Ionic detergent for denaturing proteins prior to gel electrophoresis.

Specifications

Chemical Name or Material SDS, Powder
CAS 151-21-3
Quantity 250 g
Molecular Formula C12H25NaO4S
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.