missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford SDS, Powder
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
5 kg
25 kg
Unit Size:
100g
1kg
250g
25kg
500g
5kg
Description
Ionic detergent for denaturing proteins prior to gel electrophoresis.
Ionic detergent for denaturing proteins prior to gel electrophoresis.
Specifications
Specifications
| Chemical Name or Material | SDS, Powder |
| CAS | 151-21-3 |
| Quantity | 500 g |
| Molecular Formula | C12H25NaO4S |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction