missing translation for 'onlineSavingsMsg'
Learn More

Melford SDS Pellets

Product Code. p-200087051
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy

Product Code. 30018383

Brand: Melford L220401000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

New micro-pellets allow convenient weighing and handling without the mess and odor of? powders.

New micro-pellets allow convenient weighing and handling without the mess and odor of powders. High purity SDS is additive free. Easy to dissolve in water for denaturing proteins prior to gel elecrophoresis.

Specifications

Chemical Name or Material SDS, Pellets
CAS 151-21-3
Quantity 1 kg
Molecular Formula C12H25NaO4S
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.