missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SD 1008

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
JAK2/STAT3 signaling pathway inhibitor. Inhibits activation of STAT3, JAK2 and Src. Induces apoptosis in cell lines expressing constitutively active tyrosine-phosphorylated STAT3.
Specifications
Tekniske data
| Quantity | 50 mg |
| Formula Weight | 329.35 |
| Purity | >99% |
| Molecular Weight (g/mol) | 329.352 |
| Chemical Name or Material | SD 1008 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion