missing translation for 'onlineSavingsMsg'
Learn More

BOLA Scrubber Adaptors for Bottles

Product Code. 11783269
Click to view available options
Standard Taper Joint:
76 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11783269

Brand: BOLA N166014

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Scrubber Adaptors for Bottles Suitable for bottles with thread GL 45, incl. leading-in tube (FEP), length 300 mm

  • Consisting of PTFE body with connecting nut and two lateral GL 18 threaded necks, a FEP inlet tube with a length of 300 mm and a gas distributor with finest bores.
  • Easy in- and outlet of gas by means of hard-walled tubing (e.g. PTFE) which can be connected to the threaded necks by means of BOLA Laboratory Screw Joints.
  • Elastic tubing can be connected by means of hose connectors. Inlet tube can be shortened individually.
  • The special feature: the body of the adaptor can be turned independently from the connecting nut.
  • This means, that the completely assembled adaptor can be removed and fixed on another bottle without the risk of disarranging the tubing.
  • Suitable for bottles of company Duran Group (formerly Schott, Mainz) with GL 45 and GLS 80 thread and a volume between 100 and 5000 mL.

Specifications

Standard Taper Joint 76 mm
Length (Metric) 300 mm
Autoclavable Autoclavable
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.