Learn More
3M™ Doodlebug™ Scrub Pad 8440
Product Code. 17198850
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 30
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17198850 White Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17198850 Supplier 3M™ Doodlebug™ Supplier No. 7100194291
  • ONLINE EXCLUSIVE PRICE
    244.00€
    146.46€ / Pack of 30
    Save 97.54€
    39% Off
Estimated Shipment: 10-12-2026
Safely cleans ceramic tile, glass, and fiberglass. Removes scuff marks from floors without dulling the finish.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Scrub Pad
Color White
For Use With (Application) Cleaning
Length (Metric) 117 mm